Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Décanoate de méthyle ≥99%

Décanoate de méthyle ≥99%

Flacon verre pour solides, 50 g
Catalog # A15658.18
Décanoate de méthyle ≥99%
Catalog # A15658.18
CAS Number:  110-42-9
Décanoate de méthyle ≥99%
Décanoate de méthyle ≥99%

Flacon verre pour solides, 50 g

CAS Number:  110-42-9
Price
SFr. 83.40
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 110-42-9 Molecular Formula: CH3(CH2)8CO2CH3 Formula Weight: 186.30 Flash Point: 94°(201°F) Refractive Index: 1.4250 Melting Point: -18° Boiling Point: 223-225° Beilstein Reference: 1759170 MDL No.: MFCD00009580

Synonyms:  Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate
  • CAS Number:110-42-9
  • Formula:C₁₁H₂₂O₂
  • MDL Number:MFCD00009580
  • Molecular Weight:186.29 g/mol
  • Boiling Pt:223…225 °C
  • Melting Pt:–18 °C
  • Density:0,877
  • Flash Pt:94 °C (201 °F)
  • Storage Temperature:Température ambiante
  • EINECS:203-766-6
Specifications

Specifications

  • Type de conditionnement:
    Flacon verre pour solides
  • Cdt:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.