Acide (4-hydroxyphényl)acétique ≥99%
Acide (4-hydroxyphényl)acétique ≥99%
Glass bottle for solids, 500 g
  A15018.36
Acide (4-hydroxyphényl)acétique ≥99%
  A15018.36
 :  156-38-7
Acide (4-hydroxyphényl)acétique ≥99%
Acide (4-hydroxyphényl)acétique ≥99%
Glass bottle for solids, 500 g
 :  156-38-7
SFr. 1’120.00
1 * 500 g
Purity: 99% CAS No.: 156-38-7 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 148-150° Beilstein Reference: 1448766
 :  4-Hydroxyphenylacetic Acid
  •  :156-38-7
  •  :(C₆H₄(OH))CH₂COOH
  •  :MFCD00004347
  •  :152.15 g/mol
  •  :347 °C (1013 hPa)
  •  :148…150 °C
  •  :Température ambiante
  •  :205-851-3
  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    500 g