Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
n-Décane ≥99%

n-Décane ≥99%

100 mL
Catalog # A14732.AE
Health
Flammable
n-Décane ≥99%
Catalog # A14732.AE
CAS Number:  124-18-5
n-Décane ≥99%
n-Décane ≥99%

100 mL

CAS Number:  124-18-5
Price
SFr. 75.10
Log in for availability
1 * 100 mL
Product Details & Documents

Purity: 99%
CAS No.: 124-18-5
Molecular Formula: C10H22
Formula Weight: 142.29
Flash Point: 46°(115°F)
Physical Form: Liquid
Density: 0.730
Refractive Index: 1.4115
Appearance: Colourless
Odour: Petrol-like
Melting Point: -30°
Boiling Point: 172-174°
Solubility: Soluble in alcohol and ethers
Beilstein Reference: 1696981
UN No.: 2247
MDL No.: MFCD00008954

  • CAS Number:124-18-5
  • Formula:H₃C(CH₂)₈CH₃
  • MDL Number:MFCD00008954
  • Molecular Weight:142.28 g/mol
  • Boiling Pt:172…174 °C
  • Melting Pt:–30 °C
  • Density:0,730
  • Flash Pt:46 °C (115 °F)
  • Storage Temperature:Température ambiante
  • EINECS:204-686-4
  • UN:2247
  • ADR:3,III
Specifications

Specifications

  • Cdt:
    100 mL

Specifications