Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Diacétyle ≥99%

Flacon Verre, 500 g
Catalog # A14217.36
Flammable
Toxic
Corrosive
Diacétyle ≥99%
Catalog # A14217.36
CAS Number:  431-03-8
Diacétyle ≥99%
Diacétyle ≥99%

Flacon Verre, 500 g

CAS Number:  431-03-8
Price
SFr. 225.00
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 431-03-8 Molecular Formula: C4H6O2 Formula Weight: 86.09 Flash Point: 10°(50°F) Refractive Index: 1.3940 Odour: Sweet Melting Point: -3° Boiling Point: 87-88° Beilstein Reference: 605398 Merck Reference: 14,2966 UN No.: 2346 MDL No.: MFCD00008756

Synonyms:  2,3-Butanedione
  • CAS Number:431-03-8
  • Formula:C₄H₆O₂
  • MDL Number:MFCD00008756
  • Molecular Weight:86.09 g/mol
  • Merck Index:14,2966
  • Boiling Pt:87…88 °C
  • Melting Pt:–3 °C
  • Density:0,983
  • Flash Pt:10 °C (50 °F)
  • Storage Temperature:Température ambiante
  • EINECS:207-069-8
  • UN:2346
  • ADR:3,II
Specifications

Specifications

  • Type de conditionnement:
    Flacon Verre
  • Cdt:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.