Pyrogallol ≥98%
Pyrogallol ≥98%
Glass bottle for solids, 500 g
Catalog # A13405.36
Health
Irritant
Pyrogallol ≥98%
Catalog # A13405.36
CAS Number:  87-66-1
Pyrogallol ≥98%
Pyrogallol ≥98%
Glass bottle for solids, 500 g
CAS Number:  87-66-1
Price
351.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents
Purity: 98+% CAS No.: 87-66-1 Molecular Formula: C6H6O3 Formula Weight: 126.11 Odour: Odourless Melting Point: 131-135° Boiling Point: 309° Beilstein Reference: 907431 Merck Reference: 14,8000 UN No.: 2811 MDL No.: MFCD00002192
Synonyms:  Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
  • CAS Number:87-66-1
  • Formula:C₆H₆O₃
  • MDL Number:MFCD00002192
  • Molecular Weight:126.11 g/mol
  • Merck Index:14,8000
  • Boiling Pt:309 °C
  • Melting Pt:131…135 °C
  • Density:1,45
  • Storage Temperature:Température ambiante
  • EINECS:201-762-9
  • UN:2811
  • ADR:6.1,III
Specifications
Specifications
  • Type de conditionnement:
    Glass bottle for solids
  • Cdt:
    500 g