L-Phénylalanine ≥99%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Cryst powder Appearance: White Melting Point: ca 275° dec. Beilstein Reference: 1910408 Merck Reference: 14,7271 Optical Rotation: -34° (c=2 in water) MDL No.: MFCD00064227
Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
- CAS Number:63-91-2
- Formula:C₉H₁₁NO₂
- MDL Number:MFCD00064227
- Molecular Weight:165.19 g/mol
- Merck Index:14,7271
- Boiling Pt:313 °C (1013 hPa)
- Melting Pt:∼ 275 °C (déc.)
- Density:0,754 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-568-1
Documents
Specifications
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



