L-Phénylalanine ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Cryst powder Appearance: White Melting Point: ca 275° dec. Beilstein Reference: 1910408 Merck Reference: 14,7271 Optical Rotation: -34° (c=2 in water) MDL No.: MFCD00064227
Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
- CAS Number:63-91-2
- Formula:C₉H₁₁NO₂
- MDL Number:MFCD00064227
- Molecular Weight:165.19 g/mol
- Merck Index:14,7271
- Boiling Pt:313 °C (1013 hPa)
- Melting Pt:∼ 275 °C (déc.)
- Density:0,754 g/cm³ (20 °C)
- Storage Temperature:Température ambiante
- EINECS:200-568-1
Specifications
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:25 g



