L-Phénylalanine ≥99%
Glass bottle for solids, 1.000 g
Fournisseur: Thermo Fisher Scientific
L-Phénylalanine ≥99%
N° de catalogue A13238.0B
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 63-91-2
Aller à
Product Details & Documents
Purity: 99% CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Cryst powder Appearance: White Melting Point: ca 275° dec. Beilstein Reference: 1910408 Merck Reference: 14,7271 Optical Rotation: -34° (c=2 in water) MDL No.: MFCD00064227
Synonymes: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine
- Numéro CAS:63-91-2
- Formule:C₉H₁₁NO₂
- Numéro MDL:MFCD00064227
- Poids moléculaire:165.19 g/mol
- Index Merck:14,7271
- Point d'ébullition:313 °C (1013 hPa)
- Point de fusion:∼ 275 °C (déc.)
- Densité:0,754 g/cm³ (20 °C)
- Température de stockage:Température ambiante
- EINECS:200-568-1
Documents
Certificats
Rechercher par numéro de lotOrganic Building Blocks and Intermediates
Notes d'application
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:1.000 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



