Triphénylméthane ≥98%
Glass bottle for solids, 500 g
Purity: 98% CAS No.: 519-73-3 Molecular Formula: (C6H5)3CH Formula Weight: 244.34 Melting Point: 92-94° Boiling Point: 358-359° Merck Reference: 14,9741 MDL No.: MFCD00004763
- :519-73-3
- :C₁₉H₁₆
- :MFCD00004763
- :244.34 g/mol
- :14,9741
- :358…359 °C
- :92…94 °C
- :1,010
- :Température ambiante
- :208-275-0
- Type de conditionnement:Glass bottle for solids
- Cdt:500 g
- Cat. No.:A11710.36
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



