Levodopa ≥98%
Glass bottle for solids, 25 g
Fournisseur: Thermo Fisher Scientific
Aller à
Product Details & Documents
Purity: 98+% CAS No.: 59-92-7 Molecular Formula: C9H11NO4 Formula Weight: 197.19 Odour: Odourless Melting Point: ca 295° dec. Beilstein Reference: 2215169 Merck Reference: 14,5464 MDL No.: MFCD00002598
Synonymes: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
- Numéro CAS:59-92-7
- Formule:C₉H₁₁NO₄
- Numéro MDL:MFCD00002598
- Poids moléculaire:197.19 g/mol
- Index Merck:14,5464
- Point de fusion:∼ 295 °C (déc.)
- Température de stockage:Température ambiante
- EINECS:200-445-2
Documents
Certificats
Rechercher par numéro de lotOrganic Building Blocks and Intermediates
Notes d'application
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:25 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



