Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Acrylamide 30% en solution aqueuse

Glass bottle, 500 mL
  GRBU1107.0500
 
 
Marketsource
Acrylamide 30% en solution aqueuse
  GRBU1107.0500
Marketsource
 :  79-06-1
Acrylamide 30% en solution aqueuse
Acrylamide 30% en solution aqueuse

Glass bottle, 500 mL

 :  79-06-1
 
 
   
1 * 500 mL
 
 :  Prop-2-enamide
  •  :79-06-1
  •  :C₃H₅NO
  •  :MFCD00008032
  •  :71.08 g/mol
  •  :201-173-7
  •  :3426
  •  :6.1,III
 

 

  • Type de conditionnement:
    Glass bottle
  • Cdt:
    500 mL

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.