Dissecting scissors
Arrondi/arrondi, 115 mm
N° de catalogue MART11-936-11
Fournisseur: KLS Martin Group
Aller à
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
- TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
- Straight
Spécifications
- Type:Arrondi/arrondi
- Longueur:115 mm