Agarose, MetaPhor™
Agarose, MetaPhor™
Agarose, MetaPhor™
N° de catalogue 733-1211
Fournisseur:   LONZA
Agarose, MetaPhor™
N° de catalogue 733-1211
Fournisseur:  LONZA
Agarose, MetaPhor™
Agarose, MetaPhor™
Agarose, MetaPhor™
Prix
430.00 SFr.
Connexion Pour la disponibilité
1 * 25 g
Product Details & Documents
Intermediate melting point agarose ideal for resolution of DNA fragments, PCR and RT-PCR products, differing in size by 2%, in the range of 20 to 800 bp, by submarine gel electrophoresis.
MetaPhor™ agarose gel is ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats.
Spécifications
  • Désignation:
    Agarose, MetaPhor™
  • Seq:
    01