Closures for headspace vials
Closures for headspace vials
PTFE coated silicone cap/spring, unassembled
N° de catalogue PERCB0104241
Fournisseur:   PerkinElmer
Closures for headspace vials
N° de catalogue PERCB0104241
Fournisseur:  PerkinElmer
Closures for headspace vials
PTFE coated silicone cap/spring, unassembled
Prix
189.00 SFr.
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Spécifications
  • Désignation:
    PTFE coated silicone cap/spring, unassembled
  • Seq:
    02