D-Cyclosérine
Glass bottle for solids, 1 g
N° de catalogue MOLE69907662-1G
Marketsource
Fournisseur: Molekula
Aller à
Product Details & Documents
Synonymes: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
- Numéro CAS:68-41-7
- Formule:C₃H₆N₂O₂
- Numéro MDL:MFCD00005353
- Poids moléculaire:102.09 g/mol
- Température de stockage:Congélateur
- EINECS:200-688-4
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:1 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



