Linoléate de méthyle
Glass bottle, 5 mL
ROTH7217.1
Marketsource
: cis,cis-9,12-Octadecadienoic acid methyl ester, 9,12-Octadecanoic acid methyl ester, 9Z,12Z-Octadecadienoic acid methyl ester, Linoleic acid methyl ester, (9Z,12Z)-Methyl octadeca-9,12-dienoate, Methyl cis,cis-9,12-octadecadienoate, 9Z,12Z-Oktadekadiensäuremethylester, 9,12 Methyl octadecaotenoate methyl linoleate, cis,cis-9,12-Oktadekadiensäuremethylester
- :112-63-0
- :C₁₉H₃₄O₂
- :MFCD00009534
- :294.48 g/mol
- :340 °C (1013 hPa)
- :–35 °C
- :0,89 g/cm³ (20 °C)
- :203-993-0
- Type de conditionnement:Glass bottle
- Cdt:5 mL