Agarose
Glass bottle for solids, 100 g
Catalog # ROTH2268.2
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Melting Pt:60…90 °C
- Storage Temperature:Température ambiante
- EINECS:232-731-8
Specifications
Specifications
- Type de conditionnement:Glass bottle for solids
- Cdt:100 g