Complement reagents
Rat complement
Katalognr. ROCKC205-0005
Direkt zu
Artikel gestrichen
Product Details & Documents
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.
Dokumente
C200-0005_ C201-0005_ C205-0005_ C220-0002_ C300-0050_
Technisches Datenblatt
Mehr
Technische Daten
- Désignation:Rat complement
- Seq:27
- Cat. No.:ROCKC205-0005
- Formulation:Lyophilized