Plongeurs de tubes
Tubing sinkers, fits tubing with inner Ø 8-10 mm
N° de catalogue BELAF329030104
Plongeurs de tubes
N° de catalogue BELAF329030104
Plongeurs de tubes
Plongeurs de tubes
Tubing sinkers, fits tubing with inner Ø 8-10 mm
Prix
SFr. 94.60
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These Teflon® sinkers help to keep tubing at the bottom of the container.
  • Ideal for use with pumps and dispensers
  • Hose barb ends for simple tubing connection
  • Solvent resistant and autoclavable
Spécifications
  • Désignation:
    Tubing sinkers, fits tubing with inner Ø 8-10 mm
  • Seq:
    03