Stirrer guides
Stirrer guide, with PP-insert, 29/32, — mm
N° de catalogue WITG1.470.029
Fournisseur: witeg Labortechnik
Aller à
Product Details & Documents
Stirrer guides are of different types.
With fused-on stirrer bearing.
GTS-vacuum-stirrer, with PTFE-insert and interchangeable PTFE-vacuum-seal, screw cap GL 25.
With PP-insert, 8 mm Ø and screw cap GL 25, made of melamine.
With cooling jacket and PTFE-insert, with interchangeable PTFE-vacuum seal and screw cap GL 25, made of melamine.
Spécifications
- RN:29/32
- Désignation:Stirrer guide, with PP-insert
- Ø axe:— mm