Desiccators
Sans robinet, 140 DN, 121 mm, 171x206 mm
N° de catalogue 467-0075
Fournisseur: Bel-Art Products, a Part of SP
Aller à
Product Details & Documents
Desiccators with white PP base and clear PC domed lid with large ribbed knob. Desiccant can be placed on or under the PP plate.
- Space saving, maximum interior clearance and volume with a minimal footprint
- Neoprene O-ring ensures tight seal down to 0 °C without grease, and will not be harmed if grease is accidentally applied
Informations de commande: Supplied with PP plate which is 3.2 mm thick with 3.2 mm diameter holes for air circulation.
Spécifications
- Dimension nominale:140 DN
- Type:Sans robinet
- Hauteur utile:121 mm
- Ø ext.×h:171x206 mm