Roller bottles, Nunc™ InVitro™
Surface étendue (5X), 4200 cm², 400-1000 ml
N° de catalogue 734-2695
Fournisseur: Thermo Fisher Scientific
Roller bottles, Nunc™ InVitro™
N° de catalogue 734-2695
Fournisseur: Thermo Fisher Scientific
Aller à
Product Details & Documents
PETG, sterile.
- Quick-action ergonomic closure reduces wrist stain and increases productivity
- Wide range of sizes with surface areas from 1050 to 4200 cm²
- Available in both Standard and the Expanded Surface, which enables increased cell growth and product yield without the need to purchase additional production equipment or increase labour
- Easy-to-read graduations for medium fills
- Lot number is printed on each bottle to maximise traceability
For industrial scale production of vaccines, monoclonal antibodies or pharmaceuticals. Ideal for adherent cells.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904.
Documents
Spécifications
- Surface de croissance:4200 cm²
- Volume de travail recommandé:400-1000 ml
- Désignation:Surface étendue (5X)
- Cat. No.:734-2695
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact