Pointes de distribution, ritips®
BioClean®, 25 ml, +, Ritter ripette®, Eppendorf® Multipette 4780, 25/unité
Catalog # BZYM742115
Marketsource
Supplier: BIOZYM
Jump to:
Product Details & Documents
Les pointes de 25 et 50 ml sont fournies avec un adaptateur.
Bioclean® tips are sterile and free from DNase, RNase and pyrogens.
Delivery information: The 25 ml and 50 ml tips are supplied with adapter.
Specifications
- Emballage:25/unité
- Stérile:+
- Volume:25 ml
- Seq:14
- Style de pointe:BioClean®
- Pour l'utilisation avec:Ritter ripette®, Eppendorf® Multipette 4780