Pipette tips, PreCision Line
Blue, 1000 µl, -, 1000 µl pipettes, 10 racks of 96 tips
N° de catalogue BZYM761005
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Spécifications
- Emballage:10 racks of 96 tips
- Stérile:-
- Volume:1000 µl
- Seq:18
- Style de pointe:Blue
- Pour l'utilisation avec:1000 µl pipettes