Pipette tips, PreCision Line
Pipette tips, PreCision Line
Standard, 200 µl, -, 200 µl pipettes, 10 racks of 96 tips
  BZYM760215
Marketsource
 :   BIOZYM
Pipette tips, PreCision Line
  BZYM760215
 :  BIOZYM
Marketsource
Pipette tips, PreCision Line
Standard, 200 µl, -, 200 µl pipettes, 10 racks of 96 tips
1 * 10 ST
Transparent, yellow or blue; sterile or non sterile.
  • Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
  • Autoclavable
  • Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
  • Emballage:
    10 racks of 96 tips
  • Stérile:
    -
  • Volume:
    200 µl
  • Seq:
    14
  • Style de pointe:
    Standard
  • Pour l'utilisation avec:
    200 µl pipettes