Pipette tips, PreCision Line
Standard, 200 µl, -, 200 µl pipettes, 50 bags of 1000 tips
BZYM760213
Marketsource
: BIOZYM
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
: DIN 12650
- Emballage:50 bags of 1000 tips
- Stérile:-
- Volume:200 µl
- Seq:13
- Style de pointe:Standard
- Pour l'utilisation avec:200 µl pipettes