Pipette tips, PreCision Line
Yellow, 200 µl, -, 200 µl pipettes, 10 bags of 1000 tips
N° de catalogue BZYM760201
Marketsource
Fournisseur: BIOZYM
Aller à
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
- Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
- Autoclavable
- Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
: DIN 12650
Spécifications
- Emballage:10 bags of 1000 tips
- Stérile:-
- Volume:200 µl
- Seq:09
- Style de pointe:Yellow
- Pour l'utilisation avec:200 µl pipettes