Pipette tips, PreCision Line
Pipette tips, PreCision Line
Crystal, 10 µl, +, 10 µl Eppendorf, Brand and Socorex pipettes, 10 racks of 96 tips
N° de catalogue BZYM760008
Marketsource
Fournisseur:   BIOZYM
Pipette tips, PreCision Line
N° de catalogue BZYM760008
Fournisseur:  BIOZYM
Marketsource
Pipette tips, PreCision Line
Crystal, 10 µl, +, 10 µl Eppendorf, Brand and Socorex pipettes, 10 racks of 96 tips
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 10 ST
Product Details & Documents
Transparent, yellow or blue; sterile or non sterile.
  • Free from DNase, RNase, ATP, pyrogens, additive, dispersant, wax and metals
  • Autoclavable
  • Produced under cleanroom conditions (VDI 2083 or class 1000/US-Fed. 209 H)
Certifications: DIN 12650
Spécifications
  • Emballage:
    10 racks of 96 tips
  • Stérile:
    +
  • Volume:
    10 µl
  • Seq:
    04
  • Style de pointe:
    Crystal
  • Pour l'utilisation avec:
    10 µl Eppendorf, Brand and Socorex pipettes