O-Ring seals for flat flanges
Brides plates DN 150
N° de catalogue LENZ06248015
Fournisseur: Lenz Laborglas
Aller à
Product Details & Documents
Silicone or Silicone PTFE-coated.
- Excellent chemical resistance
- Excellent temperature resistance
Seals for flat flanges LF 60-200.
Spécifications
- Pour:Brides plates DN 150
- Seq:03