TRIS solution tampon pH 7,2 (1 mol/l)
TRIS-HCL buffer solution, 1 mol/l, 7,2, Flacon Verre, 1 L
Katalognr. ROCKMB-001
Anbieter: Rockland Immunochemicals
TRIS solution tampon pH 7,2 (1 mol/l)
Katalognr. ROCKMB-001
Anbieter: Rockland Immunochemicals
CAS Nummer:
Direkt zu
Product Details & Documents
Tris is widely used as a component of buffer solutions, such as in TAE and TBE buffer, especially for solutions of nucleic acids.
These solutions were aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilized containers. The products were tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Achtung! Bitte beachten: These products ares for research use only and are not intended for therapeutic or diagnostic applications.
Dokumente
Technische Daten
Technische Daten
- Concentration:1 mol/l
- Type de conditionnement:Flacon Verre
- Cdt:1 L
- Désignation:TRIS-HCL buffer solution
- Valeur pH:7,2
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



