Human plasma
Human plasma
Human plasma in sodium heparin, sterile, 1.000 mL
N° de catalogue ROCKD519-06-1000
Fournisseur:   Rockland Immunochemicals
Human plasma
N° de catalogue ROCKD519-06-1000
Numéro CAS:  
Human plasma
Human plasma
Human plasma in sodium heparin, sterile, 1.000 mL
Numéro CAS:  
Prix
SFr. 8’870.00
Connexion Pour la disponibilité
1 * 1.000 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
Spécifications
Spécifications
  • Cdt:
    1.000 mL
  • Désignation:
    Human plasma in sodium heparin, sterile