Human plasma
Human plasma in sodium heparin, sterile, 100 mL
ROCKD519-06-0100
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
- Cdt:100 mL
- Désignation:Human plasma in sodium heparin, sterile