Human plasma
Human plasma in potassium oxalate, sterile, 500 mL
ROCKD519-08-0500
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
- Cdt:500 mL
- Désignation:Human plasma in potassium oxalate, sterile