Human plasma
Human plasma in potassium oxalate, sterile, 50 mL
Codice catalogo ROCKD519-08-0050
Nome del fornitore: Rockland Immunochemicals
Human plasma
Codice catalogo ROCKD519-08-0050
Nome del fornitore: Rockland Immunochemicals
Numero CAS:
Vai a
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
D519-03-0050_ D519-04-0050_
Scheda dati
Specifiche
Specifiche
- Cdt:50 mL
- Désignation:Human plasma in potassium oxalate, sterile
Specifiche