Human plasma
Human plasma
Human plasma in lithium heparin, sterile, 50 mL
N° de catalogue ROCKD519-07-0050
Fournisseur:   Rockland Immunochemicals
Human plasma
N° de catalogue ROCKD519-07-0050
Numéro CAS:  
Human plasma
Human plasma
Human plasma in lithium heparin, sterile, 50 mL
Numéro CAS:  
Prix
1 390.00 SFr.
Connexion Pour la disponibilité
1 * 50 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
Spécifications
Spécifications
  • Cdt:
    50 mL
  • Désignation:
    Human plasma in lithium heparin, sterile
Spécifications