Human plasma
Human plasma in alsevers, sterile, 50 mL
N° de catalogue ROCKD519-02-0050
Fournisseur: Rockland Immunochemicals
Aller à
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
D519-03-0050_ D519-04-0050_
Fiche technique
Spécifications
Spécifications
- Cdt:50 mL
- Désignation:Human plasma in alsevers, sterile