Human plasma
Human plasma
Human plasma in ACD, sterile, 100 mL
N° de catalogue ROCKD519-01-0100
Fournisseur:   Rockland Immunochemicals
Human plasma
N° de catalogue ROCKD519-01-0100
Numéro CAS:  
Human plasma
Human plasma
Human plasma in ACD, sterile, 100 mL
Numéro CAS:  
Prix
2 400.00 SFr.
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents
Plasma is obtained from non-hemolysed blood collected from healthy, fasted donors. Sterilisation is accomplished by filtration.
Documents
Spécifications
Spécifications
  • Cdt:
    100 mL
  • Désignation:
    Human plasma in ACD, sterile