Test tubes without rim, medium wall
100 mm, 16 mm
Catalog # 212-7108
Supplier: MURRAY S
Jump to:
Product Details & Documents
Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certifications: ISO 4142:2002
Specifications
- Ø ext.:16 mm
- Longueur:100 mm