Pressure Bulbs
Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 5, PVC, 102
N° de catalogue DENE4200005
Fournisseur:   DEUTSCH NEUMANN
Pressure Bulbs
N° de catalogue DENE4200005
Fournisseur:  DEUTSCH NEUMANN
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 5, PVC, 102
Prix
39.30 SFr.
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Spécifications
  • Capacité:
    102
  • Désignation:
    Atomiser bulb with pressure valve, red-brown, size 5, PVC
  • Seq:
    3