Donkey serum
Sérum d'âne, Filtré de manière stérile, Frozen
Fournisseur: Biowest
Aller à
Product Details & Documents
Donkey serum may be used as a blocking agent and controls in immunoassays. For immunohistochemistry and immunocytochemistry, donkey serum was used between 5 to 20% as a blocking agent in various studies.
Documents
Spécifications
- Format du sérum:Frozen
- Stérilité:Filtré de manière stérile
- Désignation:Sérum d'âne
- Seq:24