Acide γ-linolénique
Glass bottle for solids, 100 mg
N° de catalogue BTRN1153
Aller à
Arrêté
Product Details & Documents
Synonymes: cis,cis,cis-6,9,12-Octadecatrienoic acid, γ-Linolenic acid, (6Z,9Z,12Z)-Octadeca-6,9,12-trienoic acid, 6Z,9Z,12Z-Octadecatrienoic acid
- Numéro CAS:506-26-3
- Formule:C₁₈H₃₀O₂
- Numéro MDL:MFCD00065718
- Poids moléculaire:278.44 g/mol
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:100 mg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



