Plaques d'électroporation
Plaques d'électroporation, écart de 2 mm, 25, 125 µl
Catalog # 732-0480
Supplier: BTX
Jump to:
Product Details & Documents
Sterile multi-well electroporation plates for transfection/transformation of bacteria, yeast, insect, plant and mammalian cells.
Documents
BTX Electroporation and CRISPR
Product Literature
Specifications
- Nb de puits:25
- Volume:125 µl
- Désignation:Plaques d'électroporation, écart de 2 mm