SOLUTION STAND. COND. CLY11-D 12,64 mS/C 1 * 500 mL
Catalog # ENDR50081905
Supplier: Endress+Hauser
Frequently Bought Together
Ferroïne (Phénanthroline-1,10-fer(II) sulfate) 0.025 M en solution aqueuse
Supplier: Thermo Fisher Scientific
Methyl Linolenate, Reference Standard
Supplier: USP
Ethyl Acetate, Reference Standard
Supplier: USP
Conductivity standards, at 25 °C, Supelco®
Supplier: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
1-FLUORONAPHTHALENE 1 * 100 mg
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Hexazinone
Supplier: Merck
Analytical Volumetric Solution Zinc Sulp 1 * 1 L
Supplier: REAGECON
[EN]HYDROCORTISONE IMPURITY G 1 * 50 mg
Supplier: Merck
Olopatadine Related Compounds, Reference Standard
Supplier: USP
PROPOFOL RELATED COMPOUND B 1 * 50 mg
Supplier: Merck
CLOTHIANIDIN CERTIFIED 1 * 0,1 g
Supplier: EHRENSTORFER