Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIP-10. Anti-Human IP-10 specific antibody was purif ied by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P93BT-1
Supplier: PeproTech, Inc.
Frequently Bought Together