ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant Rat MCP-1(MCAF). Anti-Rat MCP-1(MCAF) specific antib ody was purified by affinity chromatography and then biotinylated. 1 * 1 mg
Référence PEPR500-P76BT-1
Fournisseur:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant Rat MCP-1(MCAF). Anti-Rat MCP-1(MCAF) specific antib ody was purified by affinity chromatography and then biotinylated. 1 * 1 mg
Référence PEPR500-P76BT-1
Fournisseur:  PeproTech, Inc.
Frequently Bought Together