ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hRANTES. Anti-Human RANTES specific antibody was pur ified by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P36BT-1
Supplier:  PeproTech, Inc.
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hRANTES. Anti-Human RANTES specific antibody was pur ified by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P36BT-1
Supplier:  PeproTech, Inc.
Frequently Bought Together