Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hTNF-beta. Anti-Human TNF-beta specific antibody was purified by affinity chromatography and then biotinylated. 1 * 1 mg
Référence PEPR500-P31BBT-1
Fournisseur: PeproTech, Inc.
Frequently Bought Together