ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-6. Anti-Human IL-6 specific antibody was purifie d by affinity chromatography and then biotinylated. 1 * 1 mg
  PEPR500-P26BT-1
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-6. Anti-Human IL-6 specific antibody was purifie d by affinity chromatography and then biotinylated. 1 * 1 mg
  PEPR500-P26BT-1
Frequently Bought Together