ContactUsLinkComponent
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-1alpha. Anti-Human IL-1alpha specific antibody w as purified by affinity chromatography and then biotinylated. 1 * 1 mg
  PEPR500-P21ABT-1
Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hIL-1alpha. Anti-Human IL-1alpha specific antibody w as purified by affinity chromatography and then biotinylated. 1 * 1 mg
  PEPR500-P21ABT-1
Frequently Bought Together