Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-2. Anti-Murine IL-2 specific antibody was purifi ed by affinity chromatography and then biotinylated. 1 * 1 mg
Catalog # PEPR500-P111BT-1
Supplier: PeproTech, Inc.
Frequently Bought Together